2-Amino-4-bromobenzyl alcohol

Supplier: Apollo Scientific

Synonyms: (2-Amino-4-bromophenyl)methanol

OR471584-1G OR471584-5G
APOSOR471584-1GEA 85.8 EUR
APOSOR471584-1G APOSOR471584-5G
2-Amino-4-bromobenzyl alcohol
2-Amino-4-bromobenzyl alcohol
Formula: C₇H₈BrNO
MW: 202,049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779492
CAS Number: 946122-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR