2-(1H-Pyrrol-2-yl)acetonitrile
Supplier: Apollo Scientific
OR471598-1G
OR471598-250MG
OR471598-5G
APOSOR471598-1GEA
408
EUR
APOSOR471598-1G
APOSOR471598-250MG
APOSOR471598-5G
2-(1H-Pyrrol-2-yl)acetonitrile
2-(1H-Pyrrol-2-yl)acetonitrile
Formula:
C₆H₆N₂ MW: 106,125 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09878875 CAS Number: 50551-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...