2-Amino-5-[(4-ethyl-1-piperazinyl)methyl]pyridine 95%

Supplier: Apollo Scientific

OR471684-25G OR471684-5G
APOSOR471684-25GEA 150 EUR
APOSOR471684-25G APOSOR471684-5G
2-Amino-5-[(4-ethyl-1-piperazinyl)methyl]pyridine 95%
2-Amino-5-[(4-ethyl-1-piperazinyl)methyl]pyridine
Formula: C₁₂H₂₀N₄
Storage Temperature: Ambient
MDL Number: MFCD14706690
CAS Number: 1180132-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR