1,3-Dibromo-2-methylpropane 95%

Supplier: Apollo Scientific

OR471692-1G OR471692-5G
APOSOR471692-1GEA 66.9 EUR
APOSOR471692-1G APOSOR471692-5G
1,3-Dibromo-2-methylpropane 95%
1,3-Dibromo-2-methylpropane
Formula: C₄H₈Br₂
MW: 215,914 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18909351
CAS Number: 28148-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR