4-Amino-3,5-dichloro-2-methylpyridine 97%

Supplier: Apollo Scientific

Synonyms: 3,5-Dichloro-2-methylpyridin-4-amine

OR471695-1G OR471695-250MG
APOSOR471695-1GEA 109 EUR
APOSOR471695-1G APOSOR471695-250MG
4-Amino-3,5-dichloro-2-methylpyridine 97%
4-Amino-3,5-dichloro-2-methylpyridine
Formula: C₆H₆Cl₂N₂
MW: 177,031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00160307
CAS Number: 195045-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR