Ethyl 3-(4-morpholinophenyl)-3-oxopropanoate 95%

Supplier: Apollo Scientific

OR471703-1G OR471703-5G
APOSOR471703-1GEA 387 EUR
APOSOR471703-1G APOSOR471703-5G
Ethyl 3-(4-morpholinophenyl)-3-oxopropanoate 95%
Ethyl 3-(4-morpholinophenyl)-3-oxopropanoate
Formula: C₁₅H₁₉NO₄
MW: 277,316 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11974624
CAS Number: 55356-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR