Ethyl 3-(2-methoxy-4-methylphenyl)-3-oxopropanoate 95%
Supplier: Apollo Scientific
OR471709-1G
OR471709-250MG
APOSOR471709-1GEA
710
EUR
APOSOR471709-1G
APOSOR471709-250MG
Ethyl 3-(2-methoxy-4-methylphenyl)-3-oxopropanoate 95%
Ethyl 3-(2-methoxy-4-methylphenyl)-3-oxopropanoate
Formula:
C₁₃H₁₆O₄ MW: 236,264 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31695761 CAS Number: 125041-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...