4-Bromo-5-methyl-2-(methylthio)pyrimidine 95%

Supplier: Apollo Scientific

OR471717-1G OR471717-5G
APOSOR471717-1GEA 323 EUR
APOSOR471717-1G APOSOR471717-5G
4-Bromo-5-methyl-2-(methylthio)pyrimidine 95%
4-Bromo-5-methyl-2-(methylthio)pyrimidine
Formula: C₆H₇BrN₂S
MW: 219.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28049291
CAS Number: 1823532-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR