2-(Cyclopropylmethoxy)pyrimidine-5-carbaldehyde 95%

Supplier: Apollo Scientific

OR471723-100MG OR471723-1G OR471723-250MG
APOSOR471723-100MGEA 670 EUR
APOSOR471723-100MG APOSOR471723-1G APOSOR471723-250MG
2-(Cyclopropylmethoxy)pyrimidine-5-carbaldehyde 95%
2-(Cyclopropylmethoxy)pyrimidine-5-carbaldehyde
Formula: C₉H₁₀N₂O₂
MW: 178,188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26843559
CAS Number: 1192569-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR