N-[(1S)-1-(3-Bromophenyl)ethyl]cyclopropanamine hydrochloride
Supplier: Apollo Scientific
|
Warning
|
OR475070-1G
OR475070-250MG
APOSOR475070-1GEA
183
EUR
APOSOR475070-1G
APOSOR475070-250MG
N-[(1S)-1-(3-Bromophenyl)ethyl]cyclopropanamine hydrochloride
N-[(1S)-1-(3-Bromophenyl)ethyl]cyclopropanamine hydrochloride
Formula:
C₁₁H₁₄BrN·ClH MW: 276,601 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31695676 CAS Number: 2407348-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...