2,6-Bis(tert-butyl)isonicotinic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2,6-Bis(tert-butyl)pyridine-4-carboxylic acid

OR4775-1G OR4775-250MG
APOSOR4775-1GEA 640 EUR
APOSOR4775-1G APOSOR4775-250MG
2,6-Bis(tert-butyl)isonicotinic acid
2,6-Bis(tert-butyl)isonicotinic acid
Formula: C₁₄H₂₁NO₂
MW: 235,322 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06496458
CAS Number: 191478-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR