Ethyl 4-(2-chloroethylcarbamoylamino)benzoate

Supplier: Apollo Scientific
Warning

OR47838-1G OR47838-250MG
APOSOR47838-1GEA 26.1 EUR
APOSOR47838-1G APOSOR47838-250MG
Ethyl 4-(2-chloroethylcarbamoylamino)benzoate
Ethyl 4-(2-chloroethylcarbamoylamino)benzoate
Formula: C₁₂H₁₅ClN₂O₃
MW: 270.712 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32200407
CAS Number: 13908-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR