4-Amino-3-cyanobenzoic acid 95%

Supplier: Apollo Scientific

OR47861-1G OR47861-250MG
APOSOR47861-1GEA 116 EUR
APOSOR47861-1G APOSOR47861-250MG
4-Amino-3-cyanobenzoic acid 95%
4-Amino-3-cyanobenzoic acid
Formula: C₈H₆N₂O₂
MW: 162,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09745178
CAS Number: 74192-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR