4-Methylmandelic acid

Supplier: Apollo Scientific
Danger

Synonyms: Hydroxy(4-methylphenyl)acetic acid, 4-[Carboxy(hydroxy)methyl]toluene

OR4793-500MG
APOSOR4793-500MGEA 0 EUR
APOSOR4793-500MG
4-Methylmandelic acid
4-Methylmandelic acid
Formula: C₉H₁₀O₃
MW: 166,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00961860
CAS Number: 18584-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR