2-Phenyl-imidazo[1,2-a]pyridin-3-ylamine 97%

Supplier: Apollo Scientific

OR480216-10G OR480216-1G OR480216-5G
APOSOR480216-10GEA 721 EUR
APOSOR480216-10G APOSOR480216-1G APOSOR480216-5G
2-Phenyl-imidazo[1,2-a]pyridin-3-ylamine 97%
2-Phenyl-imidazo[1,2-a]pyridin-3-ylamine
Formula: C₁₃H₁₁N₃
MW: 209,247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01033240
CAS Number: 3999-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR