(2R)-2-Amino-3-methyl-but-3-enoic acid hydrochloride 95%
Supplier: Apollo Scientific
OR480226-1G
OR480226-250MG
OR480226-500MG
APOSOR480226-1GEA
1050
EUR
APOSOR480226-1G
APOSOR480226-250MG
APOSOR480226-500MG
(2R)-2-Amino-3-methyl-but-3-enoic acid hydrochloride 95%
(2R)-2-Amino-3-methyl-but-3-enoic acid hydrochloride
Formula:
C₅H₉NO₂·ClH MW: 151,591 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03093482 CAS Number: 60103-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...