6-Nitrooxindole 95%

Supplier: Apollo Scientific

Synonyms: 6-Nitroindolin-2-one

OR480257-1G OR480257-500MG
APOSOR480257-1GEA 815 EUR
APOSOR480257-1G APOSOR480257-500MG
6-Nitrooxindole 95%
6-Nitrooxindole
Formula: C₈H₆N₂O₃
MW: 178.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835636
CAS Number: 474799-41-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR