tert-Butyl (2R,6R)-2,6-dimethylpiperazine-1-carboxylate 95%
Supplier: Apollo Scientific
Synonyms:
(2R,6R)-tert-Butyl 2,6-dimethylpiperazine-1-carboxylate
OR480275-250MG
OR480275-500MG
APOSOR480275-250MGEA
698
EUR
APOSOR480275-250MG
APOSOR480275-500MG
tert-Butyl (2R,6R)-2,6-dimethylpiperazine-1-carboxylate 95%
tert-Butyl (2R,6R)-2,6-dimethylpiperazine-1-carboxylate
Formula:
C₁₁H₂₂N₂O₂ MW: 214,305 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11973747 CAS Number: 574007-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...