3,6-Dihydro-2H-thiopyran-5-carbaldehyde 95%

Supplier: Apollo Scientific

OR480276-1G OR480276-500MG OR480276-5G
APOSOR480276-1GEA 442 EUR
APOSOR480276-1G APOSOR480276-500MG APOSOR480276-5G
3,6-Dihydro-2H-thiopyran-5-carbaldehyde 95%
3,6-Dihydro-2H-thiopyran-5-carbaldehyde
Formula: C₆H₈OS
MW: 128,192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546919
CAS Number: 30058-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR