3-Hydroxy-1-methyl-1H-pyrazole 95%

Supplier: Apollo Scientific

Synonyms: 1-Methyl-1H-pyrazol-3(2H)-one

OR480283-10G OR480283-1G OR480283-5G
APOSOR480283-10GEA 166 EUR
APOSOR480283-10G APOSOR480283-1G APOSOR480283-5G
3-Hydroxy-1-methyl-1H-pyrazole 95%
3-Hydroxy-1-methyl-1H-pyrazole
Formula: C₄H₆N₂O
MW: 98,103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21604051
CAS Number: 52867-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR