(E)-1-(3-Bromophenyl)-3-butoxyprop-2-en-1-one
Supplier: Apollo Scientific
OR480289-1G
OR480289-250MG
OR480289-500MG
APOSOR480289-1GEA
466
EUR
APOSOR480289-1G
APOSOR480289-250MG
APOSOR480289-500MG
(E)-1-(3-Bromophenyl)-3-butoxyprop-2-en-1-one
(E)-1-(3-Bromophenyl)-3-butoxyprop-2-en-1-one
Formula:
C₁₃H₁₅BrO₂ MW: 283,161 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22683245 CAS Number: 1375008-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...