(2R)-2-Amino-4-(methylamino)-4-oxo-butanoic acid

Supplier: Apollo Scientific
Warning

OR480363-1G
APOSOR480363-1GEA 203 EUR
APOSOR480363-1G
(2R)-2-Amino-4-(methylamino)-4-oxo-butanoic acid
(2R)-2-Amino-4-(methylamino)-4-oxo-butanoic acid
Formula: C₅H₁₀N₂O₃
MW: 146,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114468
CAS Number: 7175-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR