Isoquinoline-8-carbonitrile
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
8-Cyano-2-azanaphthalene, 8-Cyanoisoquinoline
OR48040-1G
OR48040-500MG
APOSOR48040-1GEA
326
EUR
APOSOR48040-1G
APOSOR48040-500MG
Isoquinoline-8-carbonitrile
Isoquinoline-8-carbonitrile
Formula:
C₁₀H₆N₂ MW: 154,168 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10699232 CAS Number: 362606-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...