2-Amino-2-(4-pentoxyphenyl)acetic acid

Supplier: Apollo Scientific

OR480453-1G OR480453-2G OR480453-5G
APOSOR480453-1GEA 171 EUR
APOSOR480453-1G APOSOR480453-2G APOSOR480453-5G
2-Amino-2-(4-pentoxyphenyl)acetic acid
2-Amino-2-(4-pentoxyphenyl)acetic acid
Formula: C₁₃H₁₉NO₃
MW: 237,295 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02662520
CAS Number: 500695-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR