1-Ethoxycarbonylcyclopropanecarboxylic acid 97%

Supplier: Apollo Scientific
Warning

OR480622-1G OR480622-5G
APOSOR480622-1GEA 0 EUR
APOSOR480622-1G APOSOR480622-5G
1-Ethoxycarbonylcyclopropanecarboxylic acid 97%
1-Ethoxycarbonylcyclopropanecarboxylic acid
Formula: C₇H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD08690114
CAS Number: 3697-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR