2-Hydroxy-4-iodo-benzaldehyde 95%
Supplier: Apollo Scientific
Synonyms:
2-Hydroxy-4-iodobenzaldehyde
OR480802-1G
APOSOR480802-1GEA
134
EUR
APOSOR480802-1G
2-Hydroxy-4-iodo-benzaldehyde 95%
2-Hydroxy-4-iodo-benzaldehyde
Formula:
C₇H₅IO₂ MW: 248,018 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08457196 CAS Number: 38170-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...