3-Bromo-5-cyclopropylaniline
Supplier: Apollo Scientific
OR48082-1G
OR48082-250MG
APOSOR48082-1GEA
633
EUR
APOSOR48082-1G
APOSOR48082-250MG
3-Bromo-5-cyclopropylaniline
3-Bromo-5-cyclopropylaniline
Formula:
C₉H₁₀BrN MW: 212,086 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD21099990 CAS Number: 1353855-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...