Oxepane-2,7-dione 92%
Supplier: Apollo Scientific
|
Warning
|
OR480837-1G
OR480837-5G
APOSOR480837-1GEA
0
EUR
APOSOR480837-1G
APOSOR480837-5G
Oxepane-2,7-dione 92%
Oxepane-2,7-dione
Formula:
C₆H₈O₃ Melting Pt: 22 °C Storage Temperature: Ambient |
MDL Number:
MFCD01593588 CAS Number: 2035-75-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...