5-(Chloromethyl)pyrimidine hydrochloride 95%

Supplier: Apollo Scientific

OR480875-1G OR480875-5G
APOSOR480875-1GEA 282 EUR
APOSOR480875-1G APOSOR480875-5G
5-(Chloromethyl)pyrimidine hydrochloride 95%
5-(Chloromethyl)pyrimidine hydrochloride
Formula: C₅H₅ClN₂·ClH
MW: 165,021 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19982294
CAS Number: 1337879-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR