3,5-Dichloroimidazo[1,2-a]pyridine-2-carboxylic acid

Supplier: Apollo Scientific
Warning

OR48094-250MG
APOSOR48094-250MGEA 169 EUR
APOSOR48094-250MG
3,5-Dichloroimidazo[1,2-a]pyridine-2-carboxylic acid
3,5-Dichloroimidazo[1,2-a]pyridine-2-carboxylic acid
Formula: C₈H₄Cl₂N₂O₂
MW: 231,036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846271
CAS Number: 1227954-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR