5-Amino-4-bromo-2-chlorophenol 95%

Supplier: Apollo Scientific
Warning

OR48111-1G OR48111-250MG
APOSOR48111-1GEA 575 EUR
APOSOR48111-1G APOSOR48111-250MG
5-Amino-4-bromo-2-chlorophenol 95%
5-Amino-4-bromo-2-chlorophenol
Formula: C₆H₅BrClNO
MW: 222,467 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27942409
CAS Number: 1232505-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR