4-Bromopyridazine hydrobromide 95%

Supplier: Apollo Scientific
Warning

OR49036-1G OR49036-250MG OR49036-5G
APOSOR49036-1GEA 215 EUR
APOSOR49036-1G APOSOR49036-250MG APOSOR49036-5G
4-Bromopyridazine hydrobromide 95%
4-Bromopyridazine hydrobromide
Formula: C₄H₃BrN₂·BrH
MW: 239.896 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15474999
CAS Number: 1220039-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR