2-Dimethylphosphorylaniline
Supplier: Apollo Scientific
Synonyms:
(2-Aminophenyl)dimethylphosphine oxide
OR49045-1G
OR49045-5G
APOSOR49045-1GEA
77.1
EUR
APOSOR49045-1G
APOSOR49045-5G
2-Dimethylphosphorylaniline
2-Dimethylphosphorylaniline
Formula:
C₈H₁₂NOP MW: 169,161 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23135529 CAS Number: 1197953-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...