DL-Tropic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Hydroxy-2-phenylpropanoic acid, Tropic acid, 3-Hydroxy-2-phenylpropionic acid, (±)-Tropic acid, 2-Phenylhydracrylic acid
OR4935-100G
OR4935-500G
OR4935-25G
APOSOR4935-100GEA
64
EUR
APOSOR4935-100G
APOSOR4935-500G
APOSOR4935-25G
DL-Tropic acid 98%
DL-Tropic acid
Formula:
C₉H₁₀O₃ MW: 166,18 g/mol Melting Pt: 116…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004255 CAS Number: 552-63-6 EINECS: 209-020-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...