(2-Amino-3-bromophenyl)methanol 95+%

Supplier: Apollo Scientific
Warning

OR500004-1G OR500004-250MG
APOSOR500004-1GEA 166 EUR
APOSOR500004-1G APOSOR500004-250MG
(2-Amino-3-bromophenyl)methanol 95+%
(2-Amino-3-bromophenyl)methanol
Formula: C₇H₈BrNO
MW: 202,049 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18389371
CAS Number: 397323-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR