1-(2-Bromopyridin-4-yl)ethanone 95+%

Supplier: Apollo Scientific
Warning

OR500017-1G OR500017-250MG
APOSOR500017-1GEA 331 EUR
APOSOR500017-1G APOSOR500017-250MG
1-(2-Bromopyridin-4-yl)ethanone 95+%
1-(2-Bromopyridin-4-yl)ethanone
Formula: C₇H₆BrNO
MW: 200,033 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07367876
CAS Number: 864674-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR