5-Bromo-2-nitrophenylacetic acid
Supplier: Apollo Scientific
|
Warning
|
OR50990-1G
OR50990-250MG
OR50990-5G
APOSOR50990-1GEA
139
EUR
APOSOR50990-1G
APOSOR50990-250MG
APOSOR50990-5G
5-Bromo-2-nitrophenylacetic acid
5-Bromo-2-nitrophenylacetic acid
Formula:
C₈H₆BrNO₄ MW: 260,042 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01096363 CAS Number: 124840-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...