5-Bromo-2-methyl-8-quinolinamine ≥98%

Supplier: Apollo Scientific

OR510122-1G OR510122-250MG
APOSOR510122-1GEA 264 EUR
APOSOR510122-1G APOSOR510122-250MG
5-Bromo-2-methyl-8-quinolinamine ≥98%
5-Bromo-2-methyl-8-quinolinamine
Formula: C₁₀H₉BrN₂
MW: 237,096 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24160463
CAS Number: 1417632-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR