1-(4-Pyren-1-ylphenyl)ethanone 98%

Supplier: Apollo Scientific

OR51030-1G OR51030-5G
APOSOR51030-1GEA 87 EUR
APOSOR51030-1G APOSOR51030-5G
1-(4-Pyren-1-ylphenyl)ethanone 98%
1-(4-Pyren-1-ylphenyl)ethanone
Formula: C₂₄H₁₆O
MW: 320,383 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30592343
CAS Number: 139111-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR