2-Iodo-6-methylbenzyl bromide
Supplier: Apollo Scientific
|
Danger
|
OR51857-1G
OR51857-250MG
OR51857-5G
APOSOR51857-1GEA
255
EUR
APOSOR51857-1G
APOSOR51857-250MG
APOSOR51857-5G
2-Iodo-6-methylbenzyl bromide
2-Iodo-6-methylbenzyl bromide
Formula:
C₈H₈BrI Storage Temperature: Ambient |
MDL Number:
MFCD18397826 CAS Number: 1261481-48-4 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...