Methyl 4-[(1S)-1-aminoethyl]benzoate 98%

Supplier: Apollo Scientific
Danger

Synonyms: (S)-4-(Methoxycarbonyl)-α-methylbenzylamine, (S)-Methyl 4-(1-aminoethyl)benzoate, (1S)-1-[4-(Methoxycarbonyl)phenyl]ethylamine

OR52064-1G OR52064-5G
APOSOR52064-1GEA 91.6 EUR
APOSOR52064-1G APOSOR52064-5G
Methyl 4-[(1S)-1-aminoethyl]benzoate 98%
Methyl 4-[(1S)-1-aminoethyl]benzoate
Formula: C₁₀H₁₃NO₂
MW: 179,216 g/mol
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD06762070
CAS Number: 222714-37-6
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR