4-Bromo-2-chloro-1H-benzimidazole

Supplier: Apollo Scientific
Warning

OR52092-1G OR52092-500MG
APOSOR52092-1GEA 1320 EUR
APOSOR52092-1G APOSOR52092-500MG
4-Bromo-2-chloro-1H-benzimidazole
4-Bromo-2-chloro-1H-benzimidazole
Formula: C₇H₄BrClN₂
MW: 231,477 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17168993
CAS Number: 1248548-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR