2-Bromoethylamine, N-CBZ protected
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Amino-2-bromoethane, N-CBZ protected, Benzyl (2-bromoethyl)carbamate
OR52103-100G
APOSOR52103-100GEA
171
EUR
APOSOR52103-100G
2-Bromoethylamine, N-CBZ protected
2-Bromoethylamine, N-CBZ protected
Formula:
C₁₀H₁₂BrNO₂ MW: 258,112 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09909490 CAS Number: 53844-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...