4,6-Dichloropyridin-2-ol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4,6-Dichloropyridin-2(1H)-one
OR52376-1G
OR52376-5G
APOSOR52376-1GEA
125
EUR
APOSOR52376-1G
APOSOR52376-5G
4,6-Dichloropyridin-2-ol
4,6-Dichloropyridin-2-ol
Formula:
C₅H₃Cl₂NO MW: 163,989 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD10698616 CAS Number: 68963-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...