Methyl 3-amino-5-methylisonicotinate 95%

Supplier: Apollo Scientific
Warning

OR52659-1G OR52659-250MG
APOSOR52659-1GEA 986 EUR
APOSOR52659-1G APOSOR52659-250MG
Methyl 3-amino-5-methylisonicotinate 95%
Methyl 3-amino-5-methylisonicotinate
Formula: C₈H₁₀N₂O₂
MW: 166,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26518249
CAS Number: 1805106-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR