Methyl 5-oxohexanoate 98%
Supplier: Apollo Scientific
OR52748-1G
OR52748-25G
OR52748-5G
APOSOR52748-1GEA
0
EUR
APOSOR52748-1G
APOSOR52748-25G
APOSOR52748-5G
Methyl 5-oxohexanoate 98%
Methyl 5-oxohexanoate
Formula:
C₇H₁₂O₃ Boiling Pt: 100…101 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038507 CAS Number: 13984-50-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...