2-(6-Bromopyridin-2-yl)acetic acid

Supplier: Apollo Scientific

OR52749-1G OR52749-250MG OR52749-5G
APOSOR52749-1GEA 91.5 EUR
APOSOR52749-1G APOSOR52749-250MG APOSOR52749-5G
2-(6-Bromopyridin-2-yl)acetic acid
2-(6-Bromopyridin-2-yl)acetic acid
Formula: C₇H₆BrNO₂
MW: 216,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10000075
CAS Number: 1093879-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR