4-Piperazin-1-ylpyridin-3-amine 95%

Supplier: Apollo Scientific

OR52775-1G OR52775-250MG
APOSOR52775-1GEA 1040 EUR
APOSOR52775-1G APOSOR52775-250MG
4-Piperazin-1-ylpyridin-3-amine 95%
4-Piperazin-1-ylpyridin-3-amine
Formula: C₉H₁₄N₄
Storage Temperature: Ambient
MDL Number: MFCD01704039
CAS Number: 78069-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR