3-Ethylsulphonylpyridine-2-sulphonamide 95%

Supplier: Apollo Scientific

Synonyms: 3-(Ethylsulphonyl)pyridine-2-sulphonamide

OR52893-1G OR52893-250MG
APOSOR52893-1GEA 127 EUR
APOSOR52893-1G APOSOR52893-250MG
3-Ethylsulphonylpyridine-2-sulphonamide 95%
3-Ethylsulphonylpyridine-2-sulphonamide
Formula: C₇H₁₀N₂O₄S₂
Storage Temperature: Ambient
MDL Number: MFCD01758695
CAS Number: 117671-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR