3-Bromo-4-(dimethylsulphamoyl)benzoic acid
Supplier: Apollo Scientific
|
Warning
|
OR52990-1G
OR52990-250MG
APOSOR52990-1GEA
205
EUR
APOSOR52990-1G
APOSOR52990-250MG
3-Bromo-4-(dimethylsulphamoyl)benzoic acid
3-Bromo-4-(dimethylsulphamoyl)benzoic acid
Formula:
C₉H₁₀BrNO₄S MW: 308,149 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11131893 CAS Number: 1019384-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...